Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134399.1 | NP_001127871.1 | 18 | 2540 | ||
NM_172522.4 | NP_766110.3 | 17 | 2447 | |||
GenBank mRNA | AK051642.1 | - | 17 | 2384 | ||
AK122555.2 | - | 19 | 2609 | |||
AK160406.1 | - | 5 | 561 | |||
BC151157.1 | - | 19 | 2570 | 9088045 | ||
BC172024.1 | - | 19 | 2570 | 9054310 |