Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134399.1 | NP_001127871.1 | 21 | 2964 | ||
NM_172522.4 | NP_766110.3 | 20 | 2871 | |||
GenBank mRNA | AK051642.1 | - | 20 | 2808 | ||
AK122555.2 | - | 22 | 3033 |