Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172523.3 | NP_766111.1 | 16 | 1970 | ||
GenBank mRNA | AK033871.1 | - | 16 | 1926 | ||
AK034368.1 | - | 11 | 3329 | |||
AK035644.1 | - | 16 | 1954 | |||
AK041274.1 | - | 16 | 1922 | |||
AK078096.1 | - | 1 | 239 | |||
BC078449.1 | - | 15 | 1841 | 5705966 |