Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172525.2 | NP_766113.1 | 3 | 404 | ||
GenBank mRNA | AK043637.1 | - | 3 | 451 | ||
AK044979.1 | - | 3 | 454 | |||
AK139908.1 | - | 1 | 361 | |||
BC040387.1 | - | 3 | 428 | 3599962 | ||
BC052858.1 | - | 3 | 428 |