Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172525.2 | NP_766113.1 | 7, 8 | 806 | ||
GenBank mRNA | AK043637.1 | - | 7, 8 | 853 | ||
AK044979.1 | - | 7, 8 | 856 | |||
AK132716.1 | - | 2, 3 | 192 | |||
AK139908.1 | - | 5, 6 | 763 | |||
BC040387.1 | - | 7, 8 | 830 | 3599962 | ||
BC052858.1 | - | 7, 8 | 830 |