Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172525.2 | NP_766113.1 | 12 | 1317 | ||
GenBank mRNA | AK043637.1 | - | 12 | 1364 | ||
AK044979.1 | - | 12 | 1367 | |||
AK132716.1 | - | 7 | 703 | |||
BC031572.1 | - | 11 | ||||
BC040387.1 | - | 12 | 1341 | 3599962 |