Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172527.2 | NP_766115.1 | 4 | 763 | ||
GenBank mRNA | AK032657.1 | - | 2 | 358 | ||
AK040749.1 | - | 4 | 766 | |||
AK043196.1 | - | 265 | ||||
AK168451.1 | - | 307 | ||||
BC060962.1 | - | 2 | 347 | 30302091 |