Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172527.2 | NP_766115.1 | 5 | 991 | ||
GenBank mRNA | AK032657.1 | - | 3 | 586 | ||
AK040749.1 | - | 5 | 994 | |||
AK043196.1 | - | 493 | ||||
AK168451.1 | - | 535 | ||||
BC060962.1 | - | 3 | 575 | 30302091 |