Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146048.1 | NP_001139520.1 | 7, 8 | 879 | ||
NM_172528.3 | NP_766116.3 | 7, 8 | 527 | |||
GenBank mRNA | AK051043.1 | - | 7, 8 | 878 | ||
AK082721.1 | - | 7, 8 | 528 | |||
AK147075.1 | - | 7, 8 | 879 | |||
AK220166.1 | - | 7, 8 | 897 | |||
BC046591.1 | - | 7, 8 | 866 | |||
BC087542.1 | - | 7, 8 | 726 | 4934980 |