Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146048.1 | NP_001139520.1 | 14 | 1750 | ||
NM_172528.3 | NP_766116.3 | 14 | 1398 | |||
GenBank mRNA | AK082721.1 | - | 14 | 1399 | ||
AK147075.1 | - | 14 | 1750 | |||
AK220166.1 | - | 14 | 1768 | |||
BC022613.1 | - | 6 | 692 | |||
BC046591.1 | - | 14 | 1737 | |||
BC087542.1 | - | 14 | 1597 | 4934980 |