Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172555.2 | NP_766143.2 | 7 | 786 | ||
GenBank mRNA | AK035810.1 | - | 7 | 769 | ||
AK042629.1 | - | 7 | 753 | |||
AK139065.1 | - | 7 | 753 | |||
BC049163.1 | - | 7 | 732 | |||
BC059271.1 | - | 7 | 780 | 2811338 | ||
DQ103505.1 | - | 7 | 592 |