Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166669.1 | NP_001160141.1 | 3 | 583 | ||
NM_001166670.1 | NP_001160142.1 | 3 | 583 | |||
NM_001166671.1 | NP_001160143.1 | 3 | 462 | |||
NM_172558.3 | NP_766146.2 | 3 | 583 | |||
GenBank mRNA | AK049216.1 | - | 3 | 518 | ||
AK131137.1 | - | 3 | 438 | |||
AK143536.1 | - | 3 | 532 | |||
BC137802.1 | - | 3 | 526 | 8860821 | ||
BC144921.1 | - | 3 | 521 | |||
BC144922.1 | - | 3 | 526 | 9053470 |