Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 16 | 3174 | ||
GenBank mRNA | AK034947.1 | - | 2 | 798 | ||
AK036710.1 | - | 15 | 2994 | |||
AK147774.1 | - | 16 | 3064 | |||
BC058512.1 | - | 16 | 2007 | 6817014 |