Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005893.2 | NP_005884.2 | 1 | 1745 | ||
GenBank mRNA | AF333334.1 | - | 1 | 1743 | ||
AK313763.1 | - | 1 | 1750 | |||
BC019251.2 | - | 1 | 1713 | 4825221 | ||
Z46967.1 | - | 1 | 1625 |