Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172568.2 | NP_766156.2 | 10, 11 | 2636 | ||
GenBank mRNA | AK077168.1 | - | 4, 5 | 1256 | ||
BC117499.1 | - | 4, 5 | 1250 |