Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167976.1 | NP_001161448.1 | 7 | 479 | ||
NM_172577.3 | NP_766165.2 | 6 | 922 | |||
GenBank mRNA | AK036988.1 | - | 6 | 662 | ||
AK041694.1 | - | 6 | 923 | |||
AK045180.1 | - | 6 | 748 | |||
AK162379.1 | - | 6 | 749 | |||
AK167535.1 | - | 6 | 630 | |||
BC057980.1 | - | 6 | 746 | 5042653 | ||
EU180236.1 | - | 7 | 479 |