Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172584.3 | NP_766172.1 | 3 | 238 | ||
GenBank mRNA | AK038913.1 | - | 3 | 239 | ||
AK209474.1 | - | 1 | 24 | |||
BC056464.1 | - | 3 | 219 | 5707377 |