Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172588.2 | NP_766176.1 | 2 | 232 | ||
GenBank mRNA | AB029501.1 | - | 2 | 121 | ||
AK037699.1 | - | 2 | 232 | |||
AK054475.1 | - | 2 | 243 | |||
AK136799.1 | - | 2 | 280 | |||
AK163675.1 | - | 2 | 197 | |||
AK165334.1 | - | 2 | 239 | |||
BC049189.1 | - | 2 | 232 | 6311617 | ||
BC062131.1 | - | 2 | 232 | 6514513 |