Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172593.3 | NP_766181.2 | 13 | 2423 | ||
GenBank mRNA | AK030747.1 | - | 13 | 2348 | ||
AK083929.1 | - | 12 | 2262 | |||
AK147427.1 | - | 13 | 2424 | |||
AK147788.1 | - | 6 | 1585 | |||
BC103780.1 | - | 12 | 2253 | 30108551 | ||
BC138127.1 | - | 12 | 2241 | 8861146 |