Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164612.1 | NP_001158084.1 | 3 | 506 | ||
NM_001164613.1 | NP_001158085.1 | 4 | 559 | |||
NM_172613.4 | NP_766201.3 | 3 | 506 | |||
GenBank mRNA | AK028776.1 | - | 4 | 561 | ||
BC048410.1 | - | 3 | 506 | 6486835 |