Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330027C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172616.2 | NP_766204.2 | 9, 10 | 1214 | ||
GenBank mRNA | AK049356.1 | - | 9, 10 | 1216 | ||
AK160639.1 | - | 9, 10 | 1200 | |||
AK173190.1 | - | 8, 9 | 952 | |||
BC137719.1 | - | 9, 10 | 1223 | 8860738 |