Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330027C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172616.2 | NP_766204.2 | 10 | 1321 | ||
GenBank mRNA | AK049356.1 | - | 10 | 1323 | ||
AK160639.1 | - | 10 | 1307 | |||
AK173190.1 | - | 9 | 1059 | |||
BC137719.1 | - | 10 | 1330 | 8860738 |