Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005909.4 | NP_005900.2 | 2, 3 | 558 | ||
GenBank mRNA | AK308147.1 | - | 2, 3 | 349 | ||
BC017240.2 | - | 2, 3 | 535 | |||
BC025240.1 | - | 2, 3 | 477 | |||
BC032516.1 | - | 2, 3 | 350 | |||
BC039822.1 | - | 2, 3 | 344 | |||
BC046114.1 | - | 2, 3 | 364 | |||
BC056145.1 | - | 2, 3 | 338 | |||
BC063669.1 | - | 2, 3 | 338 | |||
BC073993.1 | - | 2, 3 | 340 | |||
BC094834.1 | - | 2, 3 | 335 | |||
BC108733.1 | - | 2, 3 | 353 | |||
BC139918.1 | - | 2, 3 | 455 | |||
BC150196.1 | - | 2, 3 | 455 | |||
BX647077.1 | - | 2, 3 | 349 |