Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172625.2 | NP_766213.1 | 2 | 195 | ||
GenBank mRNA | AK077290.1 | - | 2 | 207 | ||
AK083718.1 | - | 2 | 196 | |||
BC060961.1 | - | 2 | 213 | 6740719 |