Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 2 | 720 | ||
NM_005916.4 | NP_005907.3 | 2 | 720 | |||
NM_182776.2 | NP_877577.1 | 2 | 611 | |||
XM_005250348.3 | XP_005250405.1 | 2 | 374 | |||
XM_017012217.1 | XP_016867706.1 | 2 | 251 | |||
GenBank mRNA | AK055379.1 | - | 2 | 588 | ||
AK096959.1 | - | 2 | 173 | |||
AK226175.1 | - | 2 | 150 | |||
AK293172.1 | - | 2 | 142 | |||
AK314575.1 | - | 2 | 187 | |||
BC009398.2 | - | 2 | 138 | 4134871 | ||
BC013375.2 | - | 2 | 156 | 3946342 | ||
D55716.1 | - | 2 | 190 | |||
D86748.1 | - | 2 | 160 | |||
KU178172.1 | - | 2 | 74 | |||
KU178173.1 | - | 2 | 74 |