Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172639.2 | NP_766227.1 | 6 | 1293 | ||
GenBank mRNA | AK077347.1 | - | 6 | 1294 |