Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164263.1 | NP_001157735.1 | 8, 9 | 760 | ||
NM_172641.3 | NP_766229.2 | 8, 9 | 760 | |||
GenBank mRNA | AK036908.1 | - | 8, 9 | 758 | ||
AK053921.1 | - | 8, 9 | 758 | |||
BC023098.1 | - | 1, 2 | 99 | |||
BC039882.1 | - | 1, 2 | 51 | |||
BC090250.1 | - | 8, 9 | 735 | 6485933 |