Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172664.3 | NP_766252.2 | 20 | 3040 | ||
GenBank mRNA | AK029773.1 | - | 19 | 2766 | ||
AK172897.1 | - | 14 | 2304 | |||
BC047088.1 | - | 1 | 386 | |||
BC051641.1 | - | 20 | 3040 | 6507968 | ||
BC057369.1 | - | 22 | 3279 |