Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242336.1 | NP_001229265.1 | 2 | 792 | ||
NM_005927.4 | NP_005918.1 | 2 | 495 | |||
GenBank mRNA | AK000358.1 | - | 1 | 455 | ||
AK315573.1 | - | 2 | 464 | |||
BC026244.1 | - | 2 | 752 | 4697443 | ||
CB994356.1 | - | 2 | 496 |