Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145919.1 | NP_001139391.1 | 5 | 2621 | ||
NM_172677.3 | NP_766265.3 | 6 | 2654 | |||
GenBank mRNA | AK039176.1 | - | 6 | 2717 | ||
AK053736.1 | - | 6 | 2689 | |||
AK135700.1 | - | 1 | 286 | |||
AK137245.1 | - | 6 | 2601 | |||
AK149731.1 | - | 1 | 200 | |||
BC022932.1 | - | 2 | 1079 | |||
BC052631.1 | - | 2 | 1971 | |||
BC057158.1 | - | 6 | 2681 | 5348717 | ||
BC067040.1 | - | 5 | 2592 | 6853196 | ||
BC067042.1 | - | 6 | 2587 | 6856248 |