Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172684.2 | NP_766272.2 | 7 | 4744 | ||
GenBank mRNA | AB101658.1 | - | 8 | 2788 | ||
AK041103.1 | - | 1 | 2047 | |||
AK042965.1 | - | 1 | 1140 | |||
AK087685.1 | - | 7 | 1752 | |||
AK122577.1 | - | 7 | 4279 | |||
AK138603.1 | - | 7 | 4288 | |||
BC094667.1 | - | 7 | 4269 |