Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075098.3 | NP_001068566.1 | 3 | 490 | ||
NM_005943.5 | NP_005934.2 | 2 | 494 | |||
NR_033233.1 | 2 | 363 | ||||
XM_011514632.2 | XP_011512934.1 | 3 | 401 | |||
XM_011514633.2 | XP_011512935.1 | 3 | 401 | |||
XM_011514634.2 | XP_011512936.1 | 2 | 298 | |||
XM_017010888.1 | XP_016866377.1 | 2 | 308 | |||
GenBank mRNA | AF034374.1 | - | 3 | 373 | ||
AJ224328.1 | - | 2 | 491 | |||
AK056740.1 | - | 2 | 309 | |||
AK091306.1 | - | 2 | 222 | |||
AK300300.1 | - | 3 | 363 | |||
BC036839.1 | - | 3 | 363 | |||
DC377054.1 | - | 3 | 496 | |||
DC410181.1 | - | 3 | 455 |