Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172693.3 | NP_766281.1 | 10 | 1684 | ||
GenBank mRNA | AB175680.1 | - | 10 | 1664 | ||
AK033638.1 | - | 10 | 1685 | |||
AK042133.1 | - | 10 | 1717 | |||
AK043977.1 | - | 9 | 1545 | |||
AK082014.1 | - | 10 | 1730 | |||
AK156751.1 | - | 10 | 1714 | |||
BC024475.1 | - | 9 | 1516 | |||
BC025639.1 | - | 9 | 1406 | |||
BC056425.1 | - | 10 | 1695 | 5716909 |