Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172701.2 | NP_766289.2 | 2, 3 | 381 | ||
GenBank mRNA | AB154826.1 | - | 2, 3 | 334 | ||
AB286960.1 | - | 3, 4 | 382 | |||
AK077467.1 | - | 2, 3 | 347 | |||
AK077909.1 | - | 2, 3 | 302 | |||
BC045150.1 | - | 2, 3 | 327 | 6309923 |