Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172701.2 | NP_766289.2 | 7, 8 | 943 | ||
GenBank mRNA | AB154826.1 | - | 7, 8 | 896 | ||
AB286960.1 | - | 8, 9 | 944 | |||
AK077467.1 | - | 7, 8 | 909 | |||
AK077909.1 | - | 7, 8 | 864 | |||
BC045150.1 | - | 7, 8 | 889 | 6309923 |