Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253386.1 | NP_001240315.1 | 7, 8 | 976 | ||
NM_172702.3 | NP_766290.2 | 7, 8 | 971 | |||
GenBank mRNA | AK075863.1 | - | 7, 8 | 939 | ||
AK158914.1 | - | 7, 8 | 977 | |||
BC031720.1 | - | 7, 8 | 952 | 4209575 | ||
CB055666.1 | - | 2, 3 | 165 |