Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005954.2 | NP_005945.1 | 3 | 385 | ||
GenBank mRNA | BC013081.1 | - | 3 | 196 | 4152899 | |
BI752092.1 | - | 373 | ||||
BI820409.1 | - | 387 | ||||
BT007030.1 | - | 3 | 149 | |||
CB155600.1 | - | 3 | 243 | |||
D13365.1 | - | 3 | 201 |