Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172722.3 | NP_766310.2 | 10 | 1043 | ||
GenBank mRNA | AK049325.1 | - | 10 | 1044 | ||
AK052060.1 | - | 10 | 1031 | |||
AK149758.1 | - | 10 | 1044 | |||
AK165175.1 | - | 10 | 1034 | |||
AK167405.1 | - | 10 | 1061 | |||
AK167491.1 | - | 10 | 1061 | |||
BC028868.1 | - | 8 | 823 | |||
BC037595.1 | - | 10 | 1024 | |||
BC055480.1 | - | 9 | 945 | |||
BC082314.1 | - | 10 | 1029 | |||
BC106182.1 | - | 10 | 1018 | |||
BC132172.1 | - | 10 | 1041 | 40130449 | ||
BC138093.1 | - | 10 | 1041 | 8861112 |