Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 9 | 1151 | ||
GenBank mRNA | AK040924.1 | - | 7 | 703 | ||
AK129135.1 | - | 6 | 809 | |||
AK146431.1 | - | 9 | 1029 | |||
AK152402.1 | - | 9 | 1029 | |||
AK153787.1 | - | 9 | 1024 | |||
AK219373.1 | - | 2 | 63 | |||
BC050232.1 | - | 7 | 748 | |||
BC057548.1 | - | 9 | 1006 | |||
BC082786.1 | - | 9 | 1124 | |||
BC099419.1 | - | 9 | 1094 | |||
BC144755.1 | - | 7 | 682 |