Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 17 | 2439 | ||
GenBank mRNA | AK040924.1 | - | 15 | 1991 | ||
AK082759.1 | - | 9 | 1266 | |||
AK129135.1 | - | 13 | 3707 | |||
AK146431.1 | - | 17 | 2317 | |||
AK152402.1 | - | 17 | 2317 | |||
AK153787.1 | - | 14 | 4132 | |||
BC057548.1 | - | 17 | 2246 | |||
BC144755.1 | - | 15 | 1922 |