Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 3, 4 | 844 | ||
GenBank mRNA | AK041667.1 | - | 3, 4 | 790 | ||
AK050733.1 | - | 3, 4 | 758 | |||
AK053801.1 | - | 3, 4 | 845 | |||
AK054218.1 | - | 3, 4 | 545 | |||
AK087531.1 | - | 3, 4 | 845 | |||
AK172614.1 | - | 3, 4 | 791 | |||
AK198635.1 | - | 1, 2 | 31 | |||
BC058940.1 | - | 2, 3 | 2923 | |||
BC079853.1 | - | 3, 4 | 778 | 5716912 | ||
BC082321.1 | - | 3, 4 | 588 | 6411746 |