Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 6 | 1868 | ||
GenBank mRNA | AK041667.1 | - | 6 | 1814 | ||
AK050733.1 | - | 6 | 1782 | |||
AK053801.1 | - | 6 | 1869 | |||
AK054218.1 | - | 6 | 1569 | |||
AK087531.1 | - | 6 | 1869 | |||
AK172614.1 | - | 6 | 1815 | |||
BC058940.1 | - | 5 | 3947 | |||
BC079853.1 | - | 6 | 1802 | 5716912 | ||
BC082321.1 | - | 6 | 1612 | 6411746 |