Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 4 | 443 | ||
NM_005962.4 | NP_005953.4 | 5 | 668 | |||
NM_130439.3 | NP_569157.2 | 5 | 866 | |||
GenBank mRNA | AK303949.1 | - | 5 | 545 | ||
AK307090.1 | - | 6 | 554 | |||
AK309718.1 | - | 5 | 481 | |||
AL538051.3 | - | 5 | 679 | |||
BC012907.1 | - | 4 | 443 | 3882557 | ||
BC035128.1 | - | 5 | 872 | 5263647 | ||
BG700036.1 | - | 5 | 652 | |||
BT007069.1 | - | 3 | 323 | |||
BX417051.2 | - | 5 | 664 | |||
CR536576.1 | - | 5 | 461 | |||
D63940.1 | - | 4 | 353 | |||
KU178207.1 | - | 3 | 323 | |||
KU178208.1 | - | 4 | 369 | |||
KU178209.1 | - | 4 | 353 |