Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172730.2 | NP_766318.2 | 9 | 983 | ||
GenBank mRNA | AK046253.1 | - | 5 | 2829 | ||
AK081505.1 | - | 8 | 3628 | |||
AK156259.1 | - | 9 | 966 | |||
BC016607.1 | - | 7 | 751 | |||
BC051147.1 | - | 9 | 977 | 6595828 |