Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172731.2 | NP_766319.2 | 20 | 4559 | ||
GenBank mRNA | AK036545.1 | - | 1 | 547 | ||
AK049341.1 | - | 7 | 1233 | |||
AK086031.1 | - | 6 | 1069 | |||
AK220153.1 | - | 21 | 5033 | |||
BC042732.1 | - | 16 | 1976 | |||
BC060664.1 | - | 20 | 4022 | |||
BC145502.1 | - | 21 | 4376 | 9054065 | ||
BC150786.1 | - | 20 | 4508 | 9087697 |