Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172734.3 | NP_766322.1 | 3, 4 | 320 | ||
GenBank mRNA | AK052288.1 | - | 3, 4 | 256 | ||
AY223819.1 | - | 3, 4 | 242 | |||
BC062170.1 | - | 2, 3 | 214 | 30135051 | ||
CJ082246.1 | - | 3, 4 | 320 |