Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172734.3 | NP_766322.1 | 14 | 2805 | ||
GenBank mRNA | AK045971.1 | - | 12 | 2542 | ||
AK052288.1 | - | 14 | 2743 | |||
AY223819.1 | - | 14 | 2727 | |||
BC038621.1 | - | 7 | 1960 |