Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931406P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172741.2 | NP_766329.1 | 1 | 784 | ||
GenBank mRNA | AK076994.1 | - | 1 | 785 | ||
AK134699.1 | - | 2 | 1185 | |||
BC060233.1 | - | 1 | 585 | 6852905 |