Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172742.2 | NP_766330.2 | 16 | 2737 | ||
GenBank mRNA | AK028584.1 | - | 4 | 1362 | ||
AK037069.1 | - | 1 | 498 | |||
AK082105.1 | - | 1 | 582 | |||
BC025088.1 | - | 1 | 799 | |||
BC041672.1 | - | 16 | 2709 | |||
BC055074.1 | - | 14 | 2401 | 30052208 | ||
BC058196.1 | - | 9 | 1839 |