Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163847.1 | NP_001157319.1 | 3 | 703 | ||
NM_001163848.1 | NP_001157320.1 | 3 | 789 | |||
NM_001163849.1 | NP_001157321.1 | 3 | 703 | |||
NM_001163850.1 | NP_001157322.1 | 3 | 703 | |||
NM_001163851.1 | NP_001157323.1 | 3 | 789 | |||
NM_001163852.1 | NP_001157324.1 | 4 | 939 | |||
NM_001163853.1 | NP_001157325.1 | 3 | 767 | |||
NM_173186.4 | NP_775278.3 | 3 | 789 | |||
GenBank mRNA | AK036088.1 | - | 3 | 656 | ||
AK045194.1 | - | 3 | 675 | |||
AK049306.1 | - | 3 | 768 | |||
AK049754.1 | - | 3 | 677 | |||
AK083833.1 | - | 3 | 675 | |||
AK084693.1 | - | 3 | 743 | |||
AK086293.1 | - | 3 | 748 | |||
AK129304.1 | - | 3 | 721 | |||
AK138431.1 | - | 3 | 664 | |||
AK147647.1 | - | 3 | 789 | |||
AK161054.1 | - | 4 | 941 | |||
BC080845.1 | - | 1 | 554 | 6850962 | ||
BC094417.1 | - | 2 | 623 | 30943242 |